Learn More
Sino Biological Anti-Porin/VDAC1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101236T44100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human VDAC1, and purified by antigen affinity chromatography.
Catalog No. 50-241-0706