Learn More
Sino Biological Human VE-Cadherin Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785376
Encompass_Preferred
The Human VE-Cadherin / CD144 ELISA Pair Set is for the quantitative determination of Human VE-Cadherin / CD144.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human VE-Cadherin / CD144 coated on a 96-well plate. Standards and samples are added to the wells, and any Human VE-Cadherin / CD144 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human VE-Cadherin / CD144 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human VE-Cadherin / CD144 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85376 Supplier Sino Biological Supplier No. SEK104335
The Human VE-Cadherin / CD144 ELISA Pair Set is for the quantitative determination of Human VE-Cadherin / CD144.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human VE-Cadherin / CD144 coated on a 96-well plate. Standards and samples are added to the wells, and any Human VE-Cadherin / CD144 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human VE-Cadherin / CD144 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human VE-Cadherin / CD144 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.