Learn More
FUJIFILM BIOSCIENCES INC Human VEGF 165 , Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 10044100UG
Encompass_Preferred
Vascular endothelial growth factor-A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways. VEGF-A has several alternatively spliced isoforms, with VEGF-165 being the most abundant. The VEGF-165 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell proliferation and angiogenesis.
Catalog No. 50-990-751