Learn More
Raybiotech Inc HUMAN VEGF-A ELISA
Supplier:  Raybiotech Inc ELHVEGF1
Encompass_Preferred
RayBio Human VEGF-A ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 10 pg/ml - 6000 pg/ml
Catalog No. 50-104-4457