Learn More
ACROBiosystems ACROBiosystems Human VEGF-D Protein, His Tag
Supplier:  ACROBiosystems VEDH52281MG
Encompass_Preferred
Recombinant Protein;1MG;Human VEGF-D, His Tag (VED-H5228) is expressed from human 293 cells (HEK293). It contains AA Phe 93 - Ser 201 (Accession # AAH27948).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.4 kDa. The protein migrates as 19-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF R3, Fc Tag (Cat. No. FL4-H5251) at 5 μg/mL (100 μL/well) can bind Human VEGF-D, His Tag (Cat. No. VED-H5228) with a linear range of 0.08-0.6 μg/mL (QC tested).
Catalog No. 50-210-6755