Learn More
Abcam Human Fibrinogen ELISA Kit - high sensitivity, 1 x 96 Tests
Supplier:  Abcam AB2413831X96TESTS
Encompass_Preferred
Human Fibrinogen (FGA) ELISA kit is a single-wash, 90-minute SimpleStep ELISA used to quantify fibrinogen levels with a sensitivity of 29 pg/mL. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric assay – 450 nm readout, compatible with any standard plate reader - Different formats for different needs – 10 x 96 well plates for bulk orders andn 384-well format available for high-throughput applications - Design your own immunoassay: we also offer the conjugation-ready antibody pair in a carrier-free format
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2603893