Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery V-PLEX Plus Human VEGF Kit, Catalog Number K151RHG-1, Analyte VEGF-A, Singleplex Immunoassay, Sensitivity 1.12 pg/mL, Dynamic Range 1.12 - 562 pg/mL, Sample Types Serum, Plasma, Urine, Cell Culture Supernatant
SDP

Catalog No. NC2075736
Encompass

The MESO SCALE DISCOVERY V-PLEX Plus Human VEGF Kit is an analytically validated immunoassay designed for the quantitative measurement of human Vascular Endothelial Growth Factor A (VEGF-A). Built on the V-PLEX platform, this kit ensures reliable and reproducible results, critical for diverse research applications. Optimized for use with various biological matrices, it provides flexibility in experimental design and is compatible with multiple imaging instruments.

  • Analyte: VEGF-A
  • Species reactivity: Human
  • Assay type: Singleplex immunoassay
  • Detection platform: V-PLEX Plus
  • Plate format: 96-well
  • Sensitivity (LLOD): 1.12 pg/mL
  • Dynamic range: 1.12 - 562 pg/mL
  • Sample types: Serum, Plasma, Urine, Cell Culture Supernatant
  • Capture antibody: Mouse Monoclonal
  • Detection antibody: Mouse Monoclonal
  • Research use only: Yes
  • Kit includes: Controls, Wash Buffer, Plate Seals

Catalog No. NC2075736 Supplier Meso Scale Discovery Supplier No. K151RHG1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MESO SCALE DISCOVERY V-PLEX Plus Human VEGF Kit is an analytically validated immunoassay designed for the quantitative measurement of human Vascular Endothelial Growth Factor A (VEGF-A). Built on the V-PLEX platform, this kit ensures reliable and reproducible results, critical for diverse research applications. Optimized for use with various biological matrices, it provides flexibility in experimental design and is compatible with multiple imaging instruments.

  • Analyte: VEGF-A
  • Species reactivity: Human
  • Assay type: Singleplex immunoassay
  • Detection platform: V-PLEX Plus
  • Plate format: 96-well
  • Sensitivity (LLOD): 1.12 pg/mL
  • Dynamic range: 1.12 - 562 pg/mL
  • Sample types: Serum, Plasma, Urine, Cell Culture Supernatant
  • Capture antibody: Mouse Monoclonal
  • Detection antibody: Mouse Monoclonal
  • Research use only: Yes
  • Kit includes: Controls, Wash Buffer, Plate Seals

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.