Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human VEGF Quantikine QuicKit ELISA

Catalog No. QK293
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QK293 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QK293 Supplier R&D Systems™ Supplier No. QK293
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Assay Range 28.10 to 1800 pg/mL
Assay Sensitivity 3.19 pg/mL
Specificity Natural and recombinant human VEGF.
For Use With (Application) Control
Gene ID (Entrez) 7422
Quantity 1 Kit
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Target Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.