Learn More
Bon Opus Biosciences Recombinant Human VEGF-A/VEGF165
SDP
Supplier:  Bon Opus Biosciences C08310UG
Encompass_Preferred
Recombinant Human Vascular Endothelial Growth Factor A is produced by our Mammalian expression system and the target gene encoding Ala27-Arg191 is expressed. Bon Opus Cat. #C083
Catalog No. 50-721-266