Learn More
ACROBiosystems ACROBiosystems Human VEGF165-coupled Magnetic Beads
Supplier:  ACROBiosystems MBSK03610MG
Encompass_Preferred
Beads;10MG;The magnetic beads technology makes use of the easy and efficient collection of beads in magnetic field to facilitate antibody purification in a simple workflow of “bind-wash-elute”. In contrast to common separation techniques, this method does not require columns or centrifugation, and is therefore ideal in high-throughput applications. ;Immobilized 31.6 μg VEGF165 protein/1mg beads can bind Anti-VEGF Antibody with an EC50 of 0.7726 μg/mL (QC tested).
Catalog No. 50-210-6310