Learn More
Abcam Human VEGFA Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502700575
Encompass_Preferred
Human VEGFA Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human VEGFA.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-0575 Supplier Abcam Supplier No. AB31286010X96TESTS
Human VEGFA Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human VEGFA.
The product is subject to the following: Abcam Restricted Use Statement