Learn More
Sino Biological VEGF121 / VEGF-A Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 10008T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VEGF121 / VEGF-A (rh VEGF121 / VEGF-A; Catalog#10008-HNAH; P15692-9; Met1-Arg147). VEGF121 / VEGF-A specific IgG was purified by Human VEGF121 / VEGF-A affinity chromatography.
Catalog No. 50-240-8755