Learn More
Sino Biological Anti-VEGF-C Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10542MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VEGF C (rh VEGF C; Catalog#10542-H08H; NP_005420.1; Thr103-Arg227). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2303