Learn More
Sino Biological Recombinant Anti-VEGF-D Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10557R106100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF-D (rh VEGF-D; Catalog#10557-H08H; NP_004460.1; Phe 93-Ser 201).
Catalog No. 50-241-1654