Learn More
Sino Biological Anti-VEGF-D Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10557RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VEGF-D (rh VEGF-D; Catalog#10557-H08H; NP_004460.1; Phe 93-Ser 201). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1636