Learn More
Sino Biological Recombinant Human VEGF-D Protein (His Tag)
Supplier:  Sino Biological 10557H08H1
Encompass_Preferred
A DNA sequence encoding the mature form of human VEGFD (NP_004460.1) corresponding to amino acid (Phe 93-Ser 201) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-255-5265