Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human VEGF121, Avi Tag (Avitag™);HEK293;25UG;VE1,RP1-261G23.1,MGC70609,MVCD1,MVCD-1,VEGF 165,VEGF-A,VEGFA,VPF,VEGF165,VEGF-165,VEGF121,VEGF120,VEGF186,VEGF164,VEGF110
Supplier:  ACROBiosystems VE1H82E725UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human VEGF121, Avi Tag (Avitag™);25UG;MABSol Biotinylated Human VEGF121, His Tag (VE1-H82E1) is expressed from human HEK293 cells. It contains AA Ala 27 - Arg 147 (Accession # P15692-9).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 16.7 kDa. As a result of glycosylation, the protein migrates as 18 kDa and 22 kDa on a SDS-PAGE gel under reducing (R) condition and 35-40 kDa under non-reducing (NR) condition.;Immobilized VEGFR1/R2-Fc at 1g/mL (100 L/well) can bind Biotinylated Human VEGF121, His Tag (Cat. No. VE1-H82E7) with a linear range of 0.1-2 ng/mL (QC tested).
Catalog No. 50-108-9756