Learn More
Invitrogen™ Human VEGFR1 Recombinant Protein
Description
RP-75749 contains purified recombinant human VeGF Receptor-1 protein with an N-terminal hexahistidine tag. RP-75749 may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P17948 |
| Concentration | 0.75 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 25 mM sodium acetate with 50% glycerol and no preservative; pH 4.8 |
| Gene ID (Entrez) | 2321 |
| Name | Human VEGFR1 |
| pH Range | 4.8 |
| Quantity | 100 μg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.