Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human VENTX Synthetic Peptide

Catalog No. PEP1120 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1120 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1120 Supplier Invitrogen™ Supplier No. PEP1120
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 16 amino acids near the center of human VENTX. PEP-1120 can be used as a blocking peptide with polyclonal antibody PA5-21006.

VENTX is a member of the Lymphoid-enhancing factor/T-cell factors (LEF1/TCF), which are a high-mobility group of transcriptional factors that play essential roles in cell fate determination during early embryogenesis and ontogenesis. VENTX is a member of the Vent family of homeodomain proteins. It is a novel LEF/TCF-associated transcriptional modulator. VENTX is predominately expressed in hematopoietic cells. VENTX may function as a transcriptional repressor and be involved in mesodermal patterning and hemapoietic stem cell maintenance, and has been suggested to have a potential role in the hematopoietic malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 27287
Species Human
Purification Method purified
Accession Number O95231
Common Name VENTX
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias hemopoietic progenitor homeobox protein VENTX2; homeobox protein VENTX; HPX42B; NA88A; VENT homeobox; VENT homeobox homolog; VENT-like homeobox 2; VENT-like homeobox protein 2; VENTX; VENTX2
Gene Symbol Ventx
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.