Learn More
Kalen Biomedical LLC Human Very Low Density Lipoprotein, 1mg
Catalog No. NC4036245
Encompass_Preferred
Human VLDL (d= <1.006 g/mL) is isolated from human plasma by ultracentrifugation. The purified VLDL is ultrafiltered through a membrane, and packaged aseptically under nitrogen. The product is provided in a solution with 154 mM NaCl, 5.6 mM Na2HPO4, 1.1 mM KH2PO4, and 0.34 mM EDTA, at pH 7.4.
Catalog No. NC4036245 Supplier Kalen Biomedical LLC Supplier No. 7701007
Human VLDL (d= <1.006 g/mL) is isolated from human plasma by ultracentrifugation. The purified VLDL is ultrafiltered through a membrane, and packaged aseptically under nitrogen. The product is provided in a solution with 154 mM NaCl, 5.6 mM Na2HPO4, 1.1 mM KH2PO4, and 0.34 mM EDTA, at pH 7.4.