Learn More
Biomatik Corporation Human Vesicular Monoamine Transporter 2 (VMAT2) ELISA Kit
Supplier:  Biomatik Corporation EKU08134
Encompass_Preferred
Human Vesicular Monoamine Transporter 2 (VMAT2) ELISA Kit for detecting VMAT2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q05940. Gene ID: 6571. Target Synonyms: SLC18A2; SVMT; SVAT; Solute Carrier Family 18 Member 2,Vesicular Monoamine; Monoamine transporter; Synaptic vesicular amine transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5591