Learn More
BIOHIPPO Human VGLL3 Microsample
SDP
Catalog No. 5000315366
Encompass
Human VGLL3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VGLL3 in serum plasma and other biological fluids It provides a detection range of 39 07-2500 pg/mL with a sensitivity of 15 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human VGLL3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human VGLL3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human VGLL3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15366 Supplier BIOHIPPO Supplier No. BHE15210619
Human VGLL3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VGLL3 in serum plasma and other biological fluids It provides a detection range of 39 07-2500 pg/mL with a sensitivity of 15 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human VGLL3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human VGLL3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human VGLL3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color