Learn More
BIORTUS USA INC Human VHL/EloB/EloC Protein, biotinylated, 100UG
Catalog No. NC2953319
Encompass_Preferred
Gene Name: VHL/ELOB/ELOC. Uniprot: P40337/Q15370/Q15369. Construct: VHL(M54-D213 end)/biotinylated-Avi-EloB(M1-K104)/EloC(M17-C112 end). Expression Host: E. coli. Purity: 90%.
Catalog No. NC2953319 Supplier BIORTUS USA INC Supplier No. BP180210TB
Gene Name: VHL/ELOB/ELOC. Uniprot: P40337/Q15370/Q15369. Construct: VHL(M54-D213 end)/biotinylated-Avi-EloB(M1-K104)/EloC(M17-C112 end). Expression Host: E. coli. Purity: 90%.