Learn More
Sino Biological Recombinant Anti-Vimentin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10028R090100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Vimentin (rh Vimentin; Catalog#10028-H08B; P08670; Met1-Glu466).
Catalog No. 50-240-8633