Learn More
Sino Biological Anti-Vimentin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10028T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Vimentin (rh Vimentin; Catalog#10028-H08B; P08670; Met1-Glu466). Vimentin specific IgG was purified by Human Vimentin affinity chromatography.
Catalog No. 50-255-5527