Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Vimentin ELISA Kit

Catalog No. EEL041
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL041 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL041 Supplier Invitrogen™ Supplier No. EEL041
Add to Cart
Add to Cart

ELISA

The Human Vimentin (VIM) ELISA quantitates Vimentin in serum, plasma, and other biological fluids The Human Vimentin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08670
Assay Range 7.81-500 ng/mL
Assay Sensitivity 4.69 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Intermediate filament
Sample Type Plasma, Serum
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; Hel113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminuscb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; Hel113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus;
Gene ID (Entrez) 7431
Gene Symbol Vim
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Vimentin
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.