Learn More
Bioworld Human Vimentin (Recombinant), 2 μg
Supplier:  Bioworld 220605141
Encompass_Preferred
Recombinant Human Vimentin Source: E.Coli Description : Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms. Formulation: Purified human Vimentin protein at 1mg/ml in 30mM Tris/HCL, pH 8, 9.5M urea, 2mM EDTA, 2mM DTT 10mM methylammonium chloride. Applications: ELISA Inhibition Assays Western Blotting Characterization: On SDS-PAGE commassie blue stained gel, the purified recombinant protein shows a band at 86kDa. Storage & Stability: Store vial at -200C to -800C. When stored at the recommended temperature, this protein is stable for 12 months. Usage: BioWORLD\\\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-612