Learn More
BIOHIPPO Human VIP Microsample
SDP
Catalog No. 5000314538
Encompass
Human VIP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VIP in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 7 82-500 pg/mL with a sensitivity of 2 65 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human VIP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human VIP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human VIP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14538 Supplier BIOHIPPO Supplier No. BHE15212235
Human VIP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VIP in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 7 82-500 pg/mL with a sensitivity of 2 65 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human VIP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human VIP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human VIP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color