Learn More
Biomatik Corporation Human Virus Induced Signaling Adapter (VISA) CLIA Kit
Supplier:  Biomatik Corporation EKU12000
Encompass_Preferred
Human Virus Induced Signaling Adapter (VISA) CLIA Kit for detecting VISA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q7Z434. Gene ID: 57506. Target Synonyms: CARDIF; IPS1; MAVS; Mitochondrial Antiviral-Signaling Protein; Interferon beta promoter stimulator protein 1; CARD Adaptor Inducing IFN-beta; NF-kappa-B-activating protein 031N. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6205