Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Visfatin ELISA Kit

Catalog No. EH482RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EH482RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH482RBX10 Supplier Invitrogen™ Supplier No. EH482RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human Visfatin quantitates human Visfatin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Visfatin.

Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43490
Assay Range 1.1-300 ng/mL
Assay Sensitivity 1.1 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1110035O14Rik; AI314458; AI480535; DKFZp666B131; EC 2.4.2.12; hypothetical protein; MGC117256; NAmPRTase; NAMPT; nampt1; nampta; NAMPTP1; Nicotinamide phosphoribosyltransferase; nicotinamide phosphoribosyltransferase 1; nicotinamide phosphoribosyltransferase a; nicotinamide phosphoribosyltransferase pseudogene 1; PBEF; Pbef1; pre-B cell-enhancing factor; pre-B-cell colony enhancing factor 1; pre-B-cell colony-enhancing factor 1; Pre-B-cell colony-enhancing factor 1 homolog; pre-B-cell colony-enhancing factor 1 precursor; VF; VISFATIN1110035O14Rik; AI314458; AI480535; DKFZp666B131; EC 2.4.2.12; hypothetical protein; MGC117256; NAmPRTase; NAMPT; nampt1; nampta; NAMPTP1; Nicotinamide phosphoribosyltransferase; nicotinamide phosphoribosyltransferase 1; nicotinamide phosphoribosyltransferase a; nicotinamide phosphoribosyltransferase pseudogene 1; PBEF; Pbef1; pre-B cell-enhancing factor; pre-B-cell colony enhancing factor 1; pre-B-cell colony-enhancing factor 1; Pre-B-cell colony-enhancing factor 1 homolog; pre-B-cell colony-enhancing factor 1 precursor; VF; VISFATIN;
Gene ID (Entrez) 10135
Gene Symbol NAMPT
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target NAMPT
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.