Learn More
Biomatik Corporation Human Visfatin (VF) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12144
Encompass_Preferred
Human Visfatin (VF) Mini sample ELISA Kit for detecting VF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43490. Gene ID: 10135. Target Synonyms: Nampt; NAmPRTase; PBEF; PBEF1; Pre-B-Cell Colony Enhancing Factor; Nicotinamide Phosphoribosyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6465