Learn More
Biomatik Corporation Human Vitamin A, Va Elisa Kit
Supplier:  Biomatik Corporation EKC31357
Encompass_Preferred
Human Vitamin A, Va Elisa Kit for detecting Vitamin A, Va in serum, plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-10ng/ml. Sensitivity: 0.467ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7193