Learn More
Biomatik Corporation Human Vitamin B12, Vb12 Elisa Kit
Supplier:  Biomatik Corporation EKC31358
Encompass_Preferred
Human Vitamin B12, Vb12 Elisa Kit for detecting Vitamin B12, Vb12 in serum, plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 1.498pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4433