Learn More
Biomatik Corporation Human Vitamin D Binding Protein (DBP) CLIA Kit
Supplier:  Biomatik Corporation EKU09479
Encompass_Preferred
Human Vitamin D Binding Protein (DBP) CLIA Kit for detecting DBP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02774. Gene ID: 2638. Target Synonyms: GC; VDBG; VDBP; Group-Specific Component; Gc-globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8511