Learn More
Gibco™ Human Vitronectin, Animal-Free Recombinant Protein, PeproTech®

Description
Recombinant Human Vitronectin is a 459 amino acid, single-chain, monomeric protein, which migrates at an apparent molecular weight of 75 kDa by SDS-PAGE under reducing conditions. The calculated molecular weight of Recombinant Human Vitronectin is 52.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Specifications
Specifications
Accession Number | P04004 |
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 7448 |
Molecular Weight (g/mol) | 52.2 kDa |
Name | Human Vitronectin, Animal-Free |
Quantity | 1 mg |
Source | HEK293 cells |
Regulatory Status | RUO |
Endotoxin Concentration | <0.1 EU/ μg |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.