Learn More
Raybiotech Inc Human Vitronectin ELISA Kit
Supplier:  Raybiotech Inc ELHVTN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.8 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human A2M, ApoA1, ApoC1, ApoC2, ApoC3, ApoE, ApoH, Artemin, BMP-6, Calbindin D, Ck beta 8-1 , CNTF, EMMPRIN, Endostatin, Fibrinogen, FLRG, GITR Ligand, GPBB, Hemoglobin, HTRA2, IL-17E, IL-1R3, Lactoferrin, Mesothelin, PAPP-A, PDECGF, PDGF Ralpha, Pepsinogen II, Periostin, Persephin, PTH, RAP, SAA, Serpin F1, Tie-1, tPA, TSP-2, Ubiquitin+1, Vitronectin, vWF
Catalog No. 50-194-9855