Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human VKORC1 Synthetic Peptide

Catalog No. PEP1401 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1401 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1401 Supplier Invitrogen™ Supplier No. PEP1401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 15 amino acids near the amino terminus of human VKORC1. PEP-1401 can be used as a blocking peptide with polyclonal antibody PA5-34358.

Vitamin K epoxide reductase complex subunit 1 (VKORC1) is the enzyme that is responsible for reducing vitamin K 2,3-epoxide to the enzymatically activated form which is essential for blood clotting. This enzymatically activated form of vitamin K is a reduced form required for the carboxylation of glutamic acid residues in some blood-clotting proteins. Fatal bleeding can be caused by vitamin K deficiency and by the vitamin K antagonist warfarin, and it is VKORC1 that is sensitive to warfarin. In humans, mutations in this gene can be associated with deficiencies in vitamin-K-dependent clotting factors and, in humans and rats, with warfarin resistance.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 79001
Species Human
Purification Method purified
Accession Number Q9BQB6
Common Name VKORC1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias D7Wsu86e; EDTP308; IMAGE3455200; MST134; MST576; MSTP134; MSTP576; phylloquinone epoxide reductase; UNQ308/PRO351; vitamin K dependent clotting factors deficiency 2; vitamin K epoxide reductase complex subunit 1; vitamin K epoxide reductase complex, subunit 1; vitamin K1 2,3-epoxide reductase subunit 1; vitamin K1 epoxide reductase (warfarin-sensitive); VKCFD2; VKOR; Vkorc1
Gene Symbol VKORC1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.