Learn More
Sino Biological Anti-VLDL Receptor/VLDLR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11075RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VLDLR (rh VLDLR; Catalog#11075-H08H; NP_003374.3; Met 1-Ser 797). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4876