Learn More
Sino Biological Human VNN2 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11728MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VNN2 / Vanin-2 (rh VNN2 / Vanin-2; Catalog#11728-H08H; NP_004656.2; Met 1-Ser 492). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3061