Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human VNN3 Synthetic Peptide

Catalog No. PEP1501 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1501 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1501 Supplier Invitrogen™ Supplier No. PEP1501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 19 amino acids near the carboxy terminus of human VNN3. PEP-1501 can be used as a blocking peptide with polyclonal antibody PA5-34459.

The vanin family is a novel group of ectoenzymes that function in tissue repair and plays a role in oxidative-stress response. As both secreted and membrane proteins, the vanin family members have been implicated as therapeutic targets in inflammatory disease. VNN3 (vascular non-inflammatory molecule 3), also known as Vanin3, is a 501 amino acid GPI-anchored amidohydrolase that is widely expressed and is found at highest levels in blood and liver. Induced by Th17 / Th1 type cytokines, VNN3 converts pantetheine into pantothenic acid. Containing one CN hydrolase domain, VNN3 is encoded by a gene that maps to human chromosome 6q23.2.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55350
Species Human
Purification Method purified
Accession Number Q8WZ42
Common Name VNN3
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias HSA238982; pantetheinase pseudogene; pantetheinase-associated gene expressed in leukocytes (PAGEL)-alpha; RGD1560609; vanin 1 pseudogene; vanin 3; vanin-3; Vascular non-inflammatory molecule 3; VNN3
Gene Symbol VNN3
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.