Learn More
Sino Biological Human Von Willebrand Factor/vWF Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786658
Encompass_Preferred
The Human VWF ELISA Pair Set is for the quantitative determination of Human VWF.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human VWF coated on a 96-well plate. Standards and samples are added to the wells, and any Human VWF present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human VWF monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human VWF present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86658 Supplier Sino Biological Supplier No. SEKA109735
The Human VWF ELISA Pair Set is for the quantitative determination of Human VWF.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human VWF coated on a 96-well plate. Standards and samples are added to the wells, and any Human VWF present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human VWF monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human VWF present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.