Learn More
BIOHIPPO Human VPS35 Microsample
SDP
Catalog No. 5000313715
Encompass
Human VPS35 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VPS35 in serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 123 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human VPS35 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human VPS35 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human VPS35 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13715 Supplier BIOHIPPO Supplier No. BHE15210689
Human VPS35 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VPS35 in serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 123 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human VPS35 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human VPS35 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human VPS35 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color