Learn More
Sino Biological Recombinant Human VSIG2 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11543H08HB100
Encompass_Preferred
A DNA sequence encoding the human VSIG2 isoform 1 (Q96IQ7-1) extracellular domain (Met1-Ala243) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4696