Learn More
Sino Biological Human VSIG4 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502372322
Encompass_Preferred
Each vial contains 100 ng of recombinant VSIG4. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended.
Catalog No. 50-237-2322 Supplier Sino Biological Supplier No. SEK1216315
Each vial contains 100 ng of recombinant VSIG4. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended.