Learn More
Sino Biological Recombinant Anti-VSIG4 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 12163R005H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human VSIG4 (rh VSIG4; Catalog#12163-H08H; Q9Y279-1; Met 1-Pro 283), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-4445