Learn More
Sino Biological Anti-VSIG4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12163MM0320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VSIG4 (rh VSIG4; Catalog#12163-H08H; Q9Y279-1; Met1-Pro283). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4447