Learn More
Sino Biological Anti-VSIG4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12163RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VSIG4 (rh VSIG4; Catalog#12163-H08H; Q9Y279-1; Met 1-Pro 283). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4442