Learn More
Sino Biological Recombinant Human VSIG4 Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 12163H02HB20
Encompass_Preferred
A DNA sequence encoding the human VSIG4 (NP_009199.1) (Met1-Pro283) was expressed with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4448