Learn More
Sino Biological Human VSIG8 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 15654H02H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human VSIG8 (NP_001013683.1)(Met1-Gly263) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3270