Learn More
Sino Biological Human Vsir MAb 100TS (Sino Biological)
Supplier:  Sino Biological 13482M017P100
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-0968